Learn More
Cayman Chemical Escin Ia, Purity: ≥98%, CAS: 123748-68-5, 5 mg
SDP
Catalog No. 502949794
Encompass_Preferred
A triterpenoid saponin with diverse biological activities; inhibits HIV-1 protease activity in a cell-free assay (IC50 = 35 µM); inhibits the invasion of, and EMT in, MDA-MB-231 cells at 2.5, 5, and 10 µM; reduces increases in vascular permeability induced by acetic acid in mice, or histamine in rats, at 100 and 200 mg/kg; inhibits increases in serum glucose levels in oral glucose-loaded rats at 100 mg/kg
Catalog No. 50-294-9794 Supplier Cayman Chemical Supplier No. 340575 MG
A triterpenoid saponin with diverse biological activities; inhibits HIV-1 protease activity in a cell-free assay (IC50 = 35 µM); inhibits the invasion of, and EMT in, MDA-MB-231 cells at 2.5, 5, and 10 µM; reduces increases in vascular permeability induced by acetic acid in mice, or histamine in rats, at 100 and 200 mg/kg; inhibits increases in serum glucose levels in oral glucose-loaded rats at 100 mg/kg