Learn More
Cayman Chemical Escin Ib, Purity: ≥98%, CAS: 26339-90-2, 5 mg
SDP
Catalog No. 502950763
Encompass_Preferred
A triterpene saponin with diverse biological activities; inhibits HIV-1 protease (IC50 = 50 µM); inhibits carrageenan-induced paw edema in rats and compound 48/80-induced scratching behavior in mice from 50-200 mg/kg; inhibits ethanol absorption and increases glucose tolerance in rats; decreases mucosal lesion size and severity in a rat model of ethanol-induced gastric ulcers from 10-50 mg/kg
Catalog No. 50-295-0763 Supplier Cayman Chemical Supplier No. 339265 MG
A triterpene saponin with diverse biological activities; inhibits HIV-1 protease (IC50 = 50 µM); inhibits carrageenan-induced paw edema in rats and compound 48/80-induced scratching behavior in mice from 50-200 mg/kg; inhibits ethanol absorption and increases glucose tolerance in rats; decreases mucosal lesion size and severity in a rat model of ethanol-induced gastric ulcers from 10-50 mg/kg