Learn More
MSC Proto Combination Wrench, 9/16 inch, 12-Point, 1 ea
Catalog No. 501919942
Encompass
This combination wrench features a 9/16 inch opening and a 15 degree offset head, designed for professional applications requiring both speed and access. It is constructed from alloy steel with a satin finish.
  • Satin finish for corrosion resistance and a slip-resistant grip
  • Twelve-point box end allows for fine indexing and improved access
  • Anti-slip design (ASD) open end
  • Box ends incorporate the TorquePlus™ system
  • Conforms to ASME B107.100 standards
  • Tether capable design
  • Overall length of 8-5/8 in
Catalog No. 50-191-9942 Supplier MSC Supplier No. 38734927
This combination wrench features a 9/16 inch opening and a 15 degree offset head, designed for professional applications requiring both speed and access. It is constructed from alloy steel with a satin finish.
  • Satin finish for corrosion resistance and a slip-resistant grip
  • Twelve-point box end allows for fine indexing and improved access
  • Anti-slip design (ASD) open end
  • Box ends incorporate the TorquePlus™ system
  • Conforms to ASME B107.100 standards
  • Tether capable design
  • Overall length of 8-5/8 in