Learn More
MSC ASD/QMS Remote Data Collection Interface, Mahr Maxum III, SPI Micrometers, 6 ft
Catalog No. 5000510698
Encompass
The Remote Data Collection Interface facilitates seamless data transfer from measurement devices to computers. It is specifically designed for compatibility with Mahr Maxum III (6 Pin) and SPI 13-700 Series IP65 micrometers.
  • Acts as an interface for data collection
  • Compatible with Mahr Maxum III (6 Pin) micrometers
  • Compatible with SPI 13-700 Series IP65 micrometers
  • Features an overall length of 6 ft
Catalog No. 50-005-10698 Supplier MSC Supplier No. 84515626
The Remote Data Collection Interface facilitates seamless data transfer from measurement devices to computers. It is specifically designed for compatibility with Mahr Maxum III (6 Pin) and SPI 13-700 Series IP65 micrometers.
  • Acts as an interface for data collection
  • Compatible with Mahr Maxum III (6 Pin) micrometers
  • Compatible with SPI 13-700 Series IP65 micrometers
  • Features an overall length of 6 ft