Learn More
MSC ASD/QMS SmartCable Gage Interface, USB, Single Input, Keyboard Output
Catalog No. 5000510766
Encompass
This SmartCable USB Gage Interface converts the output of any manufacturer's gage into USB, allowing direct connection to a PC for data collection. It features simple plug and play setup, requiring no additional software configuration, and supports both static and dynamic operating modes.
  • Converts the output of any manufacturer's gage into USB.
  • Simple plug and play setup; no software configuration required.
  • Supports static (normal) mode and dynamic (min / max / tir) mode operations.
  • Features footswitch input, LED status light, and host command operation.
  • Draws power from the PC's USB serial port or compatible systems.
  • Connects various gages directly to PCs.
  • Compatible with DP1, DP3, DP7, and Mux 10 devices.
  • Designed for harsh shop-floor environments.
  • Lifetime warranty.
Catalog No. 50-005-10766 Supplier MSC Supplier No. 84515600
This SmartCable USB Gage Interface converts the output of any manufacturer's gage into USB, allowing direct connection to a PC for data collection. It features simple plug and play setup, requiring no additional software configuration, and supports both static and dynamic operating modes.
  • Converts the output of any manufacturer's gage into USB.
  • Simple plug and play setup; no software configuration required.
  • Supports static (normal) mode and dynamic (min / max / tir) mode operations.
  • Features footswitch input, LED status light, and host command operation.
  • Draws power from the PC's USB serial port or compatible systems.
  • Connects various gages directly to PCs.
  • Compatible with DP1, DP3, DP7, and Mux 10 devices.
  • Designed for harsh shop-floor environments.
  • Lifetime warranty.