Learn More
Arrow Engineering ASE-625 Support Stand
Catalog No. NC3870498
Encompass_Preferred
The Arrow Mixing Products ASE-625 Support Stand works well with a variety of overhead stirrers with an overall height of 28″ with a 5/8″ dia center rod.
Catalog No. NC3870498 Supplier Arrow Engineering Supplier No. ASE625
The Arrow Mixing Products ASE-625 Support Stand works well with a variety of overhead stirrers with an overall height of 28″ with a 5/8″ dia center rod.