Learn More
Echelon Biosciences Research Labs a-Secretase Substrate II , Fluorogenic - 5 mg,
SDP
Catalog No. 501908679
Encompass_Preferred
CBR 6551 / a-Secretase Substrate II , Fluorogenic - 5mg , Ac-Arg-Glu(EDANS)-Val-His-His-Gln-Lys-Leu-Val-Phe-Lys(Dabcyl)-Arg-OH Category/Application: Alzheimer's This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-190-8679 Supplier Echelon Biosciences Research Labs Supplier No. 640555MG
CBR 6551 / a-Secretase Substrate II , Fluorogenic - 5mg , Ac-Arg-Glu(EDANS)-Val-His-His-Gln-Lys-Leu-Val-Phe-Lys(Dabcyl)-Arg-OH Category/Application: Alzheimer's This item is non returnable