Learn More
New England Biolabs, Inc. AseI – 10000 units
SDP
Catalog No. 50812182
Encompass
A restriction endonuclease that recognizes the sequence AT^TA_AT.
Catalog No. 50-812-182 Supplier New England Biolabs, Inc. Supplier No. R0526L
A restriction endonuclease that recognizes the sequence AT^TA_AT.