Learn More
New England Biolabs, Inc. AseI – 2000 units
SDP
Catalog No. 50812184
Encompass
A restriction endonuclease that recognizes the sequence AT^TA_AT.
Catalog No. 50-812-184 Supplier New England Biolabs, Inc. Supplier No. R0526S
A restriction endonuclease that recognizes the sequence AT^TA_AT.