Learn More
New England Biolabs, Inc. AseI – 10000 units
SDP
Supplier:  New England Biolabs, Inc. R0526M
Encompass
A restriction endonuclease that recognizes the sequence AT^TA_AT.
Catalog No. 50-812-183