Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Asenapine | 65576-45-6 | MFCD09838016 | 99.6% | 285.77 g/mol | C17H16ClNO | 25 MG
SDP

Supplier:  Medchemexpress LLC HY1012125MG

Encompass_Preferred

Asenapine is a research-grade chemical standard used in neuroscience and pharmacology research. It is an atypical antipsychotic that antagonizes serotonin, dopamine, histamine, and adrenergic receptors. Supplied as a white to off-white solid in a 25 MG vial, it is intended for laboratory research use only.

  • High purity 99.62% suitable for analytical applications.
  • Molecular formula C17H16ClNO; molecular weight 285.77 g/mol.
  • Available as a 25 MG solid vial for experimental use.
  • Storage conditions support long-term stability (powder: -20°C or 4°C).
  • Options include ready-to-use 10 mM solutions in DMSO.

Catalog No. 50-002-11486


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.