Learn More
Apexbio Technology LLC Asenapine 85650-56-2 1g
SDP
Catalog No. 5000093616
Encompass_Preferred
Asenapine (CAS 85650-56-2) is a small-molecule psychotropic agent characterized by high-affinity antagonism at multiple neurotransmitter receptors including several serotonin (5-HT) dopamine noradrenaline and histamine receptor subtypes (with reported pK sub i /sub values in the high nanomolar to sub-nanomolar range such as 8 60 8 40 10 15 9 75 and 10 46) This broad receptor antagonism profile underlies its capacity to modulate neurotransmission in various central nervous system pathways Asenapine is widely employed in biomedical research to investigate the pharmacological basis of antipsychotic activity and to study the functional roles of monoaminergic and histaminergic signaling in psychiatric and neuropharmacological models
Catalog No. 50-000-93616 Supplier Apexbio Technology LLC Supplier No. A50101000
Asenapine (CAS 85650-56-2) is a small-molecule psychotropic agent characterized by high-affinity antagonism at multiple neurotransmitter receptors including several serotonin (5-HT) dopamine noradrenaline and histamine receptor subtypes (with reported pK sub i /sub values in the high nanomolar to sub-nanomolar range such as 8 60 8 40 10 15 9 75 and 10 46) This broad receptor antagonism profile underlies its capacity to modulate neurotransmission in various central nervous system pathways Asenapine is widely employed in biomedical research to investigate the pharmacological basis of antipsychotic activity and to study the functional roles of monoaminergic and histaminergic signaling in psychiatric and neuropharmacological models