Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Asenapine hydrochloride | 1412458-61-7 | 99.95% | 322.23 | 1 ML
SDP

Catalog No. 5000393897
Encompass_Preferred

Asenapine hydrochloride (Org 5222) is an atypical antipsychotic primarily acting as an antagonist of serotonin, adrenoceptor, dopamine, and histamine receptors. It is investigated for its potential therapeutic effects in schizophrenia and bipolar disorder research, demonstrating high affinity for various 5-HT and dopamine receptor subtypes.

  • Acts as an antagonist for serotonin, adrenoceptor, dopamine, and histamine receptors.
  • Shows high affinity for 5-HT2C, 5-HT2A, 5-HT2B, 5-HT7, 5-HT6, α2B, and D3 receptors.
  • Potent antagonist at 5-HT1A, 5-HT1B, 5-HT2A, 5-HT2B, 5-HT2C, 5-HT6, 5-HT7, D2, D3, α2A, α2B, α2C, and H1 receptors.
  • Used in research for schizophrenia and bipolar disorder.
  • May offer superior therapeutic effects on anxiety symptoms.
  • Exhibits anxiolytic-like effects in mice.

Catalog No. 50-003-93897 Supplier Medchemexpress LLC Supplier No. HY1656710MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Asenapine hydrochloride (Org 5222) is an atypical antipsychotic primarily acting as an antagonist of serotonin, adrenoceptor, dopamine, and histamine receptors. It is investigated for its potential therapeutic effects in schizophrenia and bipolar disorder research, demonstrating high affinity for various 5-HT and dopamine receptor subtypes.

  • Acts as an antagonist for serotonin, adrenoceptor, dopamine, and histamine receptors.
  • Shows high affinity for 5-HT2C, 5-HT2A, 5-HT2B, 5-HT7, 5-HT6, α2B, and D3 receptors.
  • Potent antagonist at 5-HT1A, 5-HT1B, 5-HT2A, 5-HT2B, 5-HT2C, 5-HT6, 5-HT7, D2, D3, α2A, α2B, α2C, and H1 receptors.
  • Used in research for schizophrenia and bipolar disorder.
  • May offer superior therapeutic effects on anxiety symptoms.
  • Exhibits anxiolytic-like effects in mice.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.