Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Asenapine hydrochloride | 1412458-61-7 | 99.95% | 322.23 | 100 MG
SDP

Catalog No. 5000394513
Encompass_Preferred

Asenapine hydrochloride is an atypical antipsychotic that acts as an antagonist of serotonin, adreno, dopamine, and histamine receptors. It is utilized in research for schizophrenia and bipolar disorder.

  • Acts as an antagonist of serotonin receptors (pKi: 8.4-10.5).
  • Acts as an antagonist of adrenoceptors (pKi: 8.9-9.5).
  • Acts as an antagonist of dopamine receptors (pKi: 8.9-9.4).
  • Acts as an antagonist of histamine receptors (pKi: 8.2-9.0).
  • Purity of 99.95%.
  • Soluble in H2O (100 mg/mL) and DMSO (50 mg/mL).

Catalog No. 50-003-94513 Supplier Medchemexpress LLC Supplier No. HY16567100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Asenapine hydrochloride is an atypical antipsychotic that acts as an antagonist of serotonin, adreno, dopamine, and histamine receptors. It is utilized in research for schizophrenia and bipolar disorder.

  • Acts as an antagonist of serotonin receptors (pKi: 8.4-10.5).
  • Acts as an antagonist of adrenoceptors (pKi: 8.9-9.5).
  • Acts as an antagonist of dopamine receptors (pKi: 8.9-9.4).
  • Acts as an antagonist of histamine receptors (pKi: 8.2-9.0).
  • Purity of 99.95%.
  • Soluble in H2O (100 mg/mL) and DMSO (50 mg/mL).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.