Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Asenapine hydrochloride (Org 5222 hydrochloride) | 1412458-61-7 | 99.95% | 322.23 | 50 MG
SDP

Catalog No. 5000394243
Encompass_Preferred

Asenapine hydrochloride (Org 5222) is an atypical antipsychotic primarily investigated for its role in schizophrenia and bipolar disorder research. It acts as a potent antagonist across a diverse range of receptors, including serotonin, adrenoceptors, dopamine, and histamine.

  • Exhibits higher affinity for 5-HT2C, 5-HT2A, 5-HT2B, 5-HT7, 5-HT6, α2B, and D3 receptors relative to its D2 receptor affinity.
  • Potent antagonist (pKB) at 5-HT1A (7.4), 5-HT1B (8.1), 5-HT2A (9.0), 5-HT2B (9.3), 5-HT2C (9.0), 5-HT6 (8.0), 5-HT7 (8.5), D2 (9.1), D3 (9.1), α2A (7.3), α2B (8.3), α2C (6.8), and H1 (8.4) receptors.

Catalog No. 50-003-94243 Supplier Medchemexpress LLC Supplier No. HY1656750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Asenapine hydrochloride (Org 5222) is an atypical antipsychotic primarily investigated for its role in schizophrenia and bipolar disorder research. It acts as a potent antagonist across a diverse range of receptors, including serotonin, adrenoceptors, dopamine, and histamine.

  • Exhibits higher affinity for 5-HT2C, 5-HT2A, 5-HT2B, 5-HT7, 5-HT6, α2B, and D3 receptors relative to its D2 receptor affinity.
  • Potent antagonist (pKB) at 5-HT1A (7.4), 5-HT1B (8.1), 5-HT2A (9.0), 5-HT2B (9.3), 5-HT2C (9.0), 5-HT6 (8.0), 5-HT7 (8.5), D2 (9.1), D3 (9.1), α2A (7.3), α2B (8.3), α2C (6.8), and H1 (8.4) receptors.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.