Learn More
Medchemexpress LLC Asenapine maleate | 85650-56-2 | 99.95% | 401.84 | 10 MG
SDP
Catalog No. 5000343348
Encompass_Preferred
Asenapine maleate, also known by its synonym Org 5222 maleate, is a brain-penetrant atypical antipsychotic. It acts as an antagonist for various receptors, including serotonin, adrenoceptors, dopamine, and histamine receptors. It is investigated for its potential in treating schizophrenia and bipolar disorder.
  • Acts as an antagonist of serotonin receptors.
  • Exhibits antagonism towards adrenoceptors.
  • Functions as a dopamine receptor antagonist.
  • Blocks histamine receptors.
  • Investigated for its use in schizophrenia.
  • Studied for application in bipolar disorder.
  • May offer superior therapeutic effects on anxiety symptoms.
  • Demonstrates anxiolytic-like effects in behavioral tests in mice.
Catalog No. 50-003-43348 Supplier Medchemexpress LLC Supplier No. HY1110050MG
Asenapine maleate, also known by its synonym Org 5222 maleate, is a brain-penetrant atypical antipsychotic. It acts as an antagonist for various receptors, including serotonin, adrenoceptors, dopamine, and histamine receptors. It is investigated for its potential in treating schizophrenia and bipolar disorder.
  • Acts as an antagonist of serotonin receptors.
  • Exhibits antagonism towards adrenoceptors.
  • Functions as a dopamine receptor antagonist.
  • Blocks histamine receptors.
  • Investigated for its use in schizophrenia.
  • Studied for application in bipolar disorder.
  • May offer superior therapeutic effects on anxiety symptoms.
  • Demonstrates anxiolytic-like effects in behavioral tests in mice.