Learn More
Medchemexpress LLC Asenapine maleate | 85650-56-2 | 99.95% | 401.84 | 10 MG
SDP
Catalog No. 5000343654
Encompass_Preferred
Asenapine maleate (Org 5222 maleate) is a brain-penetrant atypical antipsychotic, serving as an antagonist for serotonin receptors (pKi: 8.4-10.5), adrenoceptors (pKi: 8.9-9.5), dopamine receptors (pKi: 8.9-9.4), and histamine receptors (pKi: 8.2-9.0). This compound is valuable in the research of schizophrenia and bipolar disorder.
  • Higher affinity for 5-HT2C, 5-HT2A, 5-HT2B, 5-HT7, 5-HT6, α2B, and D3 receptors compared to D2 receptor affinity.
  • Demonstrated anxiolytic-like effects in animal models.
  • Used for treating schizophrenia and bipolar I disorder.
Catalog No. 50-003-43654 Supplier Medchemexpress LLC Supplier No. HY1110010MG
Asenapine maleate (Org 5222 maleate) is a brain-penetrant atypical antipsychotic, serving as an antagonist for serotonin receptors (pKi: 8.4-10.5), adrenoceptors (pKi: 8.9-9.5), dopamine receptors (pKi: 8.9-9.4), and histamine receptors (pKi: 8.2-9.0). This compound is valuable in the research of schizophrenia and bipolar disorder.
  • Higher affinity for 5-HT2C, 5-HT2A, 5-HT2B, 5-HT7, 5-HT6, α2B, and D3 receptors compared to D2 receptor affinity.
  • Demonstrated anxiolytic-like effects in animal models.
  • Used for treating schizophrenia and bipolar I disorder.