Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Asenapine maleate | 85650-56-2 | 99.95% | 401.84 | 10 MM 1 ML
SDP

Catalog No. 5000343655
Encompass_Preferred

Asenapine maleate (Org 5222 maleate) is a brain-penetrant atypical antipsychotic. It acts as an antagonist for serotonin receptors (pKi: 8.4-10.5), adrenoceptors (pKi: 8.9-9.5), dopamine receptors (pKi: 8.9-9.4), and histamine receptors (pKi: 8.2-9.0). This compound is utilized in research for conditions such as schizophrenia and bipolar disorder.

  • Antagonist of serotonin, adrenoceptors, dopamine, and histamine receptors.
  • Brain-penetrant atypical antipsychotic.
  • Used in schizophrenia and bipolar disorder research.
  • Higher affinity for 5-HT2C, 5-HT2A, 5-HT2B, 5-HT7, 5-HT6, α2B and D3 receptors relative to D2.
  • May offer superior therapeutic effects on anxiety.
  • Demonstrates anxiolytic-like effects in animal models.

Catalog No. 50-003-43655 Supplier Medchemexpress LLC Supplier No. HY1110010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Asenapine maleate (Org 5222 maleate) is a brain-penetrant atypical antipsychotic. It acts as an antagonist for serotonin receptors (pKi: 8.4-10.5), adrenoceptors (pKi: 8.9-9.5), dopamine receptors (pKi: 8.9-9.4), and histamine receptors (pKi: 8.2-9.0). This compound is utilized in research for conditions such as schizophrenia and bipolar disorder.

  • Antagonist of serotonin, adrenoceptors, dopamine, and histamine receptors.
  • Brain-penetrant atypical antipsychotic.
  • Used in schizophrenia and bipolar disorder research.
  • Higher affinity for 5-HT2C, 5-HT2A, 5-HT2B, 5-HT7, 5-HT6, α2B and D3 receptors relative to D2.
  • May offer superior therapeutic effects on anxiety.
  • Demonstrates anxiolytic-like effects in animal models.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.