Learn More
Medchemexpress LLC Asenapine maleate | 85650-56-2 | 99.95% | 401.84 | 50 MG
SDP
Catalog No. 5000343642
Encompass_Preferred
Asenapine maleate, a brain-penetrant atypical antipsychotic, acts as an antagonist of serotonin receptors, adrenoceptors, dopamine receptors, and histamine receptors. It is used in the research of schizophrenia and bipolar disorder.
  • Exhibits high affinity for several serotonin, adrenergic, and dopamine receptors, indicating strong target engagement.
  • Acts as a potent antagonist at various serotonin, dopamine, adrenergic, and histamine receptors.
  • Demonstrates potential for superior therapeutic effect on anxiety symptoms in animal models.
  • Shows anxiolytic-like effects in behavioral tests in mice.
Catalog No. 50-003-43642 Supplier Medchemexpress LLC Supplier No. HY11100200MG
Asenapine maleate, a brain-penetrant atypical antipsychotic, acts as an antagonist of serotonin receptors, adrenoceptors, dopamine receptors, and histamine receptors. It is used in the research of schizophrenia and bipolar disorder.
  • Exhibits high affinity for several serotonin, adrenergic, and dopamine receptors, indicating strong target engagement.
  • Acts as a potent antagonist at various serotonin, dopamine, adrenergic, and histamine receptors.
  • Demonstrates potential for superior therapeutic effect on anxiety symptoms in animal models.
  • Shows anxiolytic-like effects in behavioral tests in mice.