Learn More
Medchemexpress LLC Asenapine maleate | 85650-56-2 | MFCD00900588 | 100.0% | 401.84 g/mol | C21H20ClNO5 | 5 MG
SDP
Catalog No. 5000263651
Encompass_Preferred
Asenapine maleate is the maleate salt of asenapine, an atypical antipsychotic research compound that antagonizes multiple serotonin, dopamine, histamine, and adrenergic receptors. Supplied as a high-purity solid for laboratory research use. Molecular formula C21H20ClNO5; molecular weight 401.84 g/mol; CAS 85650-56-2. Not for human or veterinary use.
  • High reported purity suitable for analytical and biochemical assays.
  • Multi-receptor antagonist profile for studies of 5-HT, D2, H1, and adrenergic signaling.
  • Solid form with defined small pack sizes for dose-response experiments.
  • Accompanied by datasheet and certificate of analysis for batch-specific quality information.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-63651 Supplier Medchemexpress LLC Supplier No. HY111005MG
Asenapine maleate is the maleate salt of asenapine, an atypical antipsychotic research compound that antagonizes multiple serotonin, dopamine, histamine, and adrenergic receptors. Supplied as a high-purity solid for laboratory research use. Molecular formula C21H20ClNO5; molecular weight 401.84 g/mol; CAS 85650-56-2. Not for human or veterinary use.
  • High reported purity suitable for analytical and biochemical assays.
  • Multi-receptor antagonist profile for studies of 5-HT, D2, H1, and adrenergic signaling.
  • Solid form with defined small pack sizes for dose-response experiments.
  • Accompanied by datasheet and certificate of analysis for batch-specific quality information.
  • Intended for research use only; not for human or veterinary use.