Learn More
Aseptic Control Products Cavicide™ Disinfectant
Catalog No. 22998800
Change view
Click to view available options
Volume (English):
24 oz.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
22-998-800 1 gal.
22-998-805 24 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-998-800 Supplier Aseptic Control Products Supplier No. 131000
Add to Cart
Add to Cart
This item is not returnable. View return policy
Specifically formulated for the control of cross-contamination
  • Use to clean equipment, laboratory surfaces, instruments, and other items that may be contaminated with infected body fluids
  • Effective against resistant organisms such as VRE and MRSA
  • Ready to use, nontoxic, and nonirritating
  • Satisfies OSHA bloodborne pathogens standards
  • Kills HIV in 3 min., turberculosis in 5 min.
  • Viricidal (hydrophilic and lipophilic)
  • Bactericidal
  • Fungicidal
Specifications
Certifications/Compliance OSHA
Composition Quaternary Ammonium and 17% Alcohol
Container Type Bottle
For Use With (Application) Used as disinfectant, cleaner and decontaminant on hard, nonporous surfaces, even in neonatal and infant care areas and most medical device matertials, effective against TB, Viruses, Bacteria, Fungi
Test Time 3 to 5 min.
Volume (English) 1 gal.