Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aseptic Control Products Cavicide™ Disinfectant

Catalog No. 22998805
Change view
Click to view available options
Volume (English):
24 oz.
1 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
22-998-805 24 oz.
22-998-800 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-998-805 Supplier Aseptic Control Products Supplier No. 131024
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifically formulated for the control of cross-contamination

  • Use to clean equipment, laboratory surfaces, instruments, and other items that may be contaminated with infected body fluids
  • Effective against resistant organisms such as VRE and MRSA
  • Ready to use, nontoxic, and nonirritating
  • Satisfies OSHA bloodborne pathogens standards
  • Kills HIV in 3 min., turberculosis in 5 min.
  • Viricidal (hydrophilic and lipophilic)
  • Bactericidal
  • Fungicidal

Specifications

Certifications/Compliance OSHA
Composition Quaternary Ammonium and 17% Alcohol
Container Type Bottle
For Use With (Application) Used as disinfectant, cleaner and decontaminant on hard, nonporous surfaces, even in neonatal and infant care areas and most medical device matertials, effective against TB, Viruses, Bacteria, Fungi
Test Time 3 to 5 min.
Volume (English) 24 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.