Learn More
Aseptic Control Products PAP Test Lubricating Jellys
Catalog No. 22998003
Change view
Click to view available options
Quantity:
12/Pk.
144/Pk.
1728/Pk.
72/Pk.
Packaging:
Packet
Tube
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
22-998-003 1728/Pk. Packet
22-998-004 144/Pk. Packet
22-998-005 72/Pk. Tube
22-998-006 12/Pk. Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Keep out of eyes and ears. Keep out of reach of children.
Catalog No. 22-998-003 Supplier Aseptic Control Products Supplier No. 024PKT
Add to Cart
Add to Cart
Keep out of eyes and ears. Keep out of reach of children.
Safe and effective for use with Pap tests
  • Eases insertions of diagnotic and therapeutic devices
  • Does not contain carbomers
  • FDA 510(k) clearance
  • Transparent and greaseless
  • Tested and approved for use with ThinPrep™ Pap Test by Hologic with Authorized Letter
Specifications
Packaging Packet
Quantity 1728/Pk.