Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aseptic Control Products VioNex™ Liquid Soap

Catalog No. p-3239092
Change view
Click to view available options
Volume (Metric):
532 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
5004345B 532 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5004345B Supplier Aseptic Control Products Supplier No. 101518

missing translation for 'validMainframeMsg'

Creamy antimicrobial liquid soap, ideal for frequent use

  • Effective germ reduction in 30 seconds
  • Lotion-based formula for extra softness
  • Mild and nondrying to skin
  • Contains emollients and softeners
  • Active ingredient: chloroxylenol (PCMX)

Developed for healthcare professionals

Specifications

Product Type Anti-microbial Soap
Form Liquid
Composition Chloroxylenol (PCMX)
For Use With (Application) Germ reduction in 30 sec.
Quantity 12 Pk.
Volume (English) 18 oz.
Volume (Metric) 532 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.