Learn More
Chemglass Life Sciences Filling Bell, Aseptic, 104mm ID, 130mm OAH
SDP
Catalog No. 501215069
Encompass_Preferred
Filling Bell, Aseptic, 104mm ID, 130mm OAH. For aseptic filling of vessels. Tubing is attached to the serrated hose connection while the receiving vessel is placed inside the bell. Made from Type 1 Class A borosilicate glass.
Catalog No. 50-121-5069 Supplier Chemglass Life Sciences Supplier No. CLS118754
Filling Bell, Aseptic, 104mm ID, 130mm OAH. For aseptic filling of vessels. Tubing is attached to the serrated hose connection while the receiving vessel is placed inside the bell. Made from Type 1 Class A borosilicate glass.