Learn More
Bellco Glass, Inc. Aseptic Fill Bell,20mm
SDP
Catalog No. 501945678
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Ideal for use of filling other glass vessels. Receiving vessel is placed under bell to maintain aseptic conditions during filling. Attaches to reservoir with rubber tubing. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassDimensions: 22x75mmTubing Size: 1/4″Size: 20mm
Catalog No. 50-194-5678 Supplier Bellco Glass, Inc. Supplier No. 561000020
Manufactured from Type 1 Class A Borosilicate Glass. Ideal for use of filling other glass vessels. Receiving vessel is placed under bell to maintain aseptic conditions during filling. Attaches to reservoir with rubber tubing. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassDimensions: 22x75mmTubing Size: 1/4″Size: 20mm