Learn More
Bellco Glass, Inc. Aseptic Fill Bell,40mm
SDP
Catalog No. 501945679
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Ideal for use of filling other glass vessels. Receiving vessel is placed under bell to maintain aseptic conditions during filling. Attaches to reservoir with rubber tubing. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassDimensions: 41x95mmTubing Size: 1/4″2 per case
Catalog No. 50-194-5679 Supplier Bellco Glass, Inc. Supplier No. 561000040
Manufactured from Type 1 Class A Borosilicate Glass. Ideal for use of filling other glass vessels. Receiving vessel is placed under bell to maintain aseptic conditions during filling. Attaches to reservoir with rubber tubing. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassDimensions: 41x95mmTubing Size: 1/4″2 per case