Learn More
Chemglass Life Sciences Filling Bell, Aseptic, 49mm ID, 100mm OAH
SDP
Catalog No. 501215067
Encompass_Preferred
Filling Bell, Aseptic, 49mm ID, 100mm OAH. For aseptic filling of vessels. Tubing is attached to the serrated hose connection while the receiving vessel is placed inside the bell. Made from Type 1 Class A borosilicate glass.
Catalog No. 50-121-5067 Supplier Chemglass Life Sciences Supplier No. CLS118752
Filling Bell, Aseptic, 49mm ID, 100mm OAH. For aseptic filling of vessels. Tubing is attached to the serrated hose connection while the receiving vessel is placed inside the bell. Made from Type 1 Class A borosilicate glass.