Learn More
Bellco Glass, Inc. Aseptic Fill Bell,70mm
SDP
Catalog No. 501945681
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Ideal for use of filling other glass vessels. Receiving vessel is placed under bell to maintain aseptic conditions during filling. Attaches to reservoir with rubber tubing. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassDimensions: 70x125mmTubing Size: 1/2″2 per case
Catalog No. 50-194-5681 Supplier Bellco Glass, Inc. Supplier No. 561000070
Manufactured from Type 1 Class A Borosilicate Glass. Ideal for use of filling other glass vessels. Receiving vessel is placed under bell to maintain aseptic conditions during filling. Attaches to reservoir with rubber tubing. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassDimensions: 70x125mmTubing Size: 1/2″2 per case