Learn More
Chemglass Life Sciences Filling Bell, Aseptic, 70mm ID, 125mm OAH
SDP
Supplier:  Chemglass Life Sciences CLS118753
Encompass_Preferred
Filling Bell, Aseptic, 70mm ID, 125mm OAH. For aseptic filling of vessels. Tubing is attached to the serrated hose connection while the receiving vessel is placed inside the bell. Made from Type 1 Class A borosilicate glass.
Catalog No. 50-121-5068