Learn More
Bellco Glass, Inc. Aseptic Sample System,50mL Bioreactor
SDP
Catalog No. 501946426
Encompass_Preferred
The aseptic sample system attaches to the headplate of a metric bioreactor allowing the user to take repeated samples into a standard 50ml disposable conical tube. The sample assembly requires an existing sample pipe (not included) which connects to the inlet pipe with tubing. The device mounts easily under a closure nut of a vessel head plate stud. Samples are taken by evacuating air with the supplied syringe and filter, thereby drawing culture media into the 50mL disposable tube.. MADE IN THE USA
Catalog No. 50-194-6426 Supplier Bellco Glass, Inc. Supplier No. 790900012
The aseptic sample system attaches to the headplate of a metric bioreactor allowing the user to take repeated samples into a standard 50ml disposable conical tube. The sample assembly requires an existing sample pipe (not included) which connects to the inlet pipe with tubing. The device mounts easily under a closure nut of a vessel head plate stud. Samples are taken by evacuating air with the supplied syringe and filter, thereby drawing culture media into the 50mL disposable tube.. MADE IN THE USA