Learn More
AVANTOR MASTERFLEX (VWR LLC) Sterilized Sampling Bottle Assembly, 250 mL
Catalog No. NC0957929
Encompass
Sterilized Sampling Bottle Assembly, 250 mL, Safely transfer, sample, and store biopharmaceutical products and reagents in applications from research to processSecure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connectionsAssemblies are pre-sterilized by gamma irradiation¿eliminates labor and validation costs associated with assembly and cleaningStores up to 250 mL
Catalog No. NC0957929 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9564402
Sterilized Sampling Bottle Assembly, 250 mL, Safely transfer, sample, and store biopharmaceutical products and reagents in applications from research to processSecure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connectionsAssemblies are pre-sterilized by gamma irradiation¿eliminates labor and validation costs associated with assembly and cleaningStores up to 250 mL