Learn More
AVANTOR MASTERFLEX (VWR LLC) Sterilized Sampling Bottle Assembly, 2 L
Catalog No. NC0957930
Encompass
Sterilized Sampling Bottle Assembly, 2 L, Safely transfer, sample, and store biopharmaceutical products and reagents in applications from research to processSecure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connectionsAssemblies are pre-sterilized by gamma irradiation¿eliminates labor and validation costs associated with assembly and cleaningStores up to 2 L
This item is not returnable. View return policy
Catalog No. NC0957930 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9564408
Sterilized Sampling Bottle Assembly, 2 L, Safely transfer, sample, and store biopharmaceutical products and reagents in applications from research to processSecure, single-piece cap/tubing assembly eliminates contamination and product loss due to loose connectionsAssemblies are pre-sterilized by gamma irradiation¿eliminates labor and validation costs associated with assembly and cleaningStores up to 2 L