Learn More
Qosina Corp AseptiQuik Cuff, Structural Support, BPA-Free, N/A
Catalog No. NC2281558
Encompass
The QOSINA AQGCUFF is a connector cuff designed for the AseptiQuik G Series, offering vital structural support in biopharmaceutical applications. It enhances fluid transfer system integrity by addressing issues like sideload and pump stress. Its genderless design allows for easy, sterile connections in various environments through a simple Flip-Click-Pull process, making it BPA-free and eliminating the need for extra clamps or welding tools.
This item is not returnable. View return policy
Catalog No. NC2281558 Supplier Qosina Corp Supplier No. AQGCUFF
The QOSINA AQGCUFF is a connector cuff designed for the AseptiQuik G Series, offering vital structural support in biopharmaceutical applications. It enhances fluid transfer system integrity by addressing issues like sideload and pump stress. Its genderless design allows for easy, sterile connections in various environments through a simple Flip-Click-Pull process, making it BPA-free and eliminating the need for extra clamps or welding tools.