Learn More
COLDER PRODUCTS COMPANY CPC Genderless AseptiQuik G Connector with MPC Insert Autoclavable Autoclavable Overall Length 1 5 in Overall Length 38 1 mm Material Silicone
Catalog No. 503131455
Encompass_Preferred
CPC Genderless AseptiQuik G Connector with MPC Insert Autoclavable Autoclavable Overall Length 1 5 in Overall Length 38 1 mm Material Silicone
Catalog No. 50-313-1455 Supplier COLDER PRODUCTS COMPANY Supplier No. AQG17MPCHT
CPC Genderless AseptiQuik G Connector with MPC Insert Autoclavable Autoclavable Overall Length 1 5 in Overall Length 38 1 mm Material Silicone