Learn More
AVANTOR MASTERFLEX (VWR LLC) CPC (Colder) Aseptiquik Standard Insert, 3/8"" Hose Barb, High Temperature
Catalog No. NC1657012
Encompass
Manufactured from sturdy polycarbonate, these connectors come presterilized and are for single-use and sanitary applications. Use connectors with filters, tubing, and process containers to create systems for upstream or downstream processes. Simply fit the connector half in your tubing line. Push the male and female connectors together; an audible snap confirms a tight connection. Align and snap membrane connectors together; pull from connector. Twist blue lock ring clockwise to hear another audible click. Alignment of the ring rib with the body¿s arrow indicator confirms final, secure connection.
Catalog No. NC1657012 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX2000295
Manufactured from sturdy polycarbonate, these connectors come presterilized and are for single-use and sanitary applications. Use connectors with filters, tubing, and process containers to create systems for upstream or downstream processes. Simply fit the connector half in your tubing line. Push the male and female connectors together; an audible snap confirms a tight connection. Align and snap membrane connectors together; pull from connector. Twist blue lock ring clockwise to hear another audible click. Alignment of the ring rib with the body¿s arrow indicator confirms final, secure connection.