Learn More
COLDER PRODUCTS COMPANY ASEPTIQUIK S SERIES
Supplier: COLDER PRODUCTS COMPANY AQS17006HT
Colder Products Company AseptiQuik S Connector High Temperature Hose Barb 3/8 in Polycarbonate Silicone platinum-cured USP Class VI ADCF Maximum Pressure 60 psi Max Operating Temperature 104 deg F 40 deg C Min Operating Temperature 39 deg F 4 deg C