Learn More
Aviva Systems Biology Corp ASF1A Antibody - N-terminal region (ARP58260_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58260_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASF1A. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ASF1A. Alternate names: CGI-98, CIA, DKFZp547E2110, HSPC146ApplicationWB. Species reactivity: Cow, Dog, Goat, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-457