Learn More
Sino Biological ASGPR1 / ASGR1 Antibody (FITC), Mouse MAb 25Tests
Supplier:  Sino Biological 10773MM02F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ASGPR1 / ASGR1 (rh ASGPR1 / ASGR1; Catalog#10773-H07H; P07306; Gln62-Leu291) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-224-8352