Learn More
Santa Cruz Biotechnology ASGPR1/Asialoglycoprotein Receptor 1/ASGR1 Antibody (8D7) PE, 200 µg/ml
Catalog No. NC3960638
Encompass
ASGPR1/Asialoglycoprotein Receptor 1/ASGR1 Antibody (8D7) is a mouse monoclonal IgG1 κ, cited in 15 publications, provided at 200 µg/mlraised against liver plasma membrane extracts of rat originAnti-ASGPR1/Asialoglycoprotein Receptor 1/ASGR1 Antibody (8D7) is recommended for detection of ASGPR1 of mouse, rat and human origin by WB, IP, IF and FCMAnti-ASGPR1/Asialoglycoprotein Receptor 1/ASGR1 Antibody (8D7) is available conjugated to agarose for IP; HRP for WB, IHC(P) and ELISA; and to either phycoerythrin or FITC for IF, IHC(P) and FCM
Catalog No. NC3960638 Supplier Santa Cruz Biotechnology Supplier No. SC52623 PE
ASGPR1/Asialoglycoprotein Receptor 1/ASGR1 Antibody (8D7) is a mouse monoclonal IgG1 κ, cited in 15 publications, provided at 200 µg/mlraised against liver plasma membrane extracts of rat originAnti-ASGPR1/Asialoglycoprotein Receptor 1/ASGR1 Antibody (8D7) is recommended for detection of ASGPR1 of mouse, rat and human origin by WB, IP, IF and FCMAnti-ASGPR1/Asialoglycoprotein Receptor 1/ASGR1 Antibody (8D7) is available conjugated to agarose for IP; HRP for WB, IHC(P) and ELISA; and to either phycoerythrin or FITC for IF, IHC(P) and FCM