Learn More
Santa Cruz Biotechnology ASGPR2/Asialoglycoprotein Receptor 2/ASGR2 Antibody (B-4) 200 µg/ml
Catalog No. NC3900875
Encompass
ASGPR2/Asialoglycoprotein Receptor 2/ASGR2 Antibody (B-4) is a mouse monoclonal IgG1 κ, cited in 1 publications, provided at 200 µg/mlraised against amino acids 81-135 mapping within an internal region of ASGPR2 of human originASGPR2/Asialoglycoprotein Receptor 2/ASGR2 Antibody (B-4) is recommended for detection of ASGPR2 of human origin by WB, IP, IF, IHC(P) and ELISA
Catalog No. NC3900875 Supplier Santa Cruz Biotechnology Supplier No. SC377113
ASGPR2/Asialoglycoprotein Receptor 2/ASGR2 Antibody (B-4) is a mouse monoclonal IgG1 κ, cited in 1 publications, provided at 200 µg/mlraised against amino acids 81-135 mapping within an internal region of ASGPR2 of human originASGPR2/Asialoglycoprotein Receptor 2/ASGR2 Antibody (B-4) is recommended for detection of ASGPR2 of human origin by WB, IP, IF, IHC(P) and ELISA