Learn More
Aviva Systems Biology Corp ASGR1 Antibody - middle region (ARP33805_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33805_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against ASGR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ASGR1. Alternate names: HL-1, ASGPR, ASGPR1, CLEC4H1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Yeast
Catalog No. 50-014-52929