Learn More
Aviva Systems Biology Corp ASGR1 Antibody - middle region (ARP33805_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33805_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASGR1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of Human ASGR1. Alternate names: ASGPR, CLEC4H1, Hs.12056, HL-1, ASGPR1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Yeast
Catalog No. 50-014-52928