Learn More
Aviva Systems Biology Corp ASGR2 Antibody - N-terminal region (ARP33820_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33820_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASGR2. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ASGR2. Alternate names: HL-2, HBXBP, ASGPR2, ASGP-R2, CLEC4H2ApplicationIHC, WB. Species reactivity: Human, Pig, Rat, Yeast
Catalog No. 50-014-55160