Learn More
Sino Biological ASGR2 / ASGPR2 Antibody (PE), Rabbit MAb 25Tests
Supplier:  Sino Biological 13908R002P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ASGR2 / ASGPR2 (rh ASGR2 / ASGPR2; Catalog#13908-H07H; P07307-1; Gln80-Ala311) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-224-8260