Learn More
Cayman Chemical ASH1L bromodomaInhumnrec 1mg
SDP
Catalog No. 502971093
Encompass_Preferred
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • MW: 42 kDa
Catalog No. 50-297-1093 Supplier Cayman Chemical Supplier No. 144891 MG
Source: Recombinant N-terminal GST-tagged protein expressed in E. coli • MW: 42 kDa