Learn More
Aviva Systems Biology Corp ASH2L Antibody - N-terminal region (ARP58134_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58134_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASH2L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ASH2L. Alternate names: ASH2, ASH2L1, ASH2L2, Bre2ApplicationWB. Species reactivity: Dog, Horse, Human, Mouse
Catalog No. 50-014-62247