Learn More
Aviva Systems Biology Corp ASH2L Antibody - middle region (ARP34202_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34202_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ASH2L. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ASH2L. Alternate names: ASH2, ASH2L1, ASH2L2, Bre2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52942