Learn More
Cole-Parmer Ashcroft 20323.0 Digital Sanitary Gauge, 1.5" Tri-Clamp, 0 to 30 psi, battery powered
Catalog No. NC1476705 Shop All Cole-Parmer Products
Encompass_Preferred
Digital sanitary gauge is designed for the pharmaceutical and biotech applications with its ±0.25% accuracy. Gauge has unlimited deadband throughout its range and superior set-point flexibility. Loop- and line-powered gauges have output signals that enable monitoring and control. Line-powered gauges offer switching capabilities.
Catalog No. NC1476705 Supplier Cole-Parmer Supplier No. 6890151
Digital sanitary gauge is designed for the pharmaceutical and biotech applications with its ±0.25% accuracy. Gauge has unlimited deadband throughout its range and superior set-point flexibility. Loop- and line-powered gauges have output signals that enable monitoring and control. Line-powered gauges offer switching capabilities.