Learn More
Grainger ASHCROFT Industrial Vacuum Gauge, 30" Hg
Catalog No. NC2835337
Encompass_Preferred
The Ashcroft 30-33HT06 Industrial Vacuum Gauge is a high-quality pressure gauge used to measure vacuum pressure. It has a 4.5 inch dial with a range of 0-30 inches of mercury. The gauge has a stainless steel case and wetted parts, with an accuracy of ±3% of full scale. It has a bottom connection of 1/4 inch NPT male. This heavy duty industrial gauge is ideal for HVAC/R, chemical processing, and other vacuum applications. The Ashcroft brand is known for exceptional quality and performance. This vacuum gauge would be an excellent addition to monitor vacuum pressure in industrial processes.
Catalog No. NC2835337 Supplier Grainger Supplier No. 33HT38
The Ashcroft 30-33HT06 Industrial Vacuum Gauge is a high-quality pressure gauge used to measure vacuum pressure. It has a 4.5 inch dial with a range of 0-30 inches of mercury. The gauge has a stainless steel case and wetted parts, with an accuracy of ±3% of full scale. It has a bottom connection of 1/4 inch NPT male. This heavy duty industrial gauge is ideal for HVAC/R, chemical processing, and other vacuum applications. The Ashcroft brand is known for exceptional quality and performance. This vacuum gauge would be an excellent addition to monitor vacuum pressure in industrial processes.